missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL 24 Protein
A cDNA sequence encoding the IL 24 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10705-1mg
Additional Details : Weight : 0.01000kg
Specifications
11009 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Measured by its ability to bind to the cell receptor of Capan-1 cells line resulted in Stat-3 activation. The ED50 for this effect is typically 1.0 ng/ml, corresponding to a Specific Activity of 1x106 units/mg. | |
Human | |
Untagged | |
Lyophilized from a 0.2microm filtered solution in PBS with BSA as a carrier. |
IL 24 Protein | |
Research Use Only | |
IL24 | |
Recombinant Protein | |
S. cerevisiae | |
MNFQQRLQSL WTLARPFCPP LLATASQMQM VVLPCLGFTL LLWSQVSGAQ GQEFHFGPCQ VKGVVPQKLW EAFWAVKDTM QAQDNITSAR LLQQEVLQNV SDAESCYLVH TLLEFYLKTV FKNYHNRTVE VRTLKSFSTL ANNFVLIVSQ LQPSQENEMF SIRDSAHRRF LLFRRAFKQL DVEAALTKAL GEVDILLTWM QKFYKL | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |