missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-13 Protein
Click to view available options
Quantity:
10 μg
100 μg
25 μg
Unit Size:
100µg
10µg
25µg
Description
The Recombinant Human IL-13 Protein is derived from E. coli. The Recombinant Human IL-13 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | AAK53823 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2um filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 3596 |
| Molecular Weight (g/mol) | 12.3 kDa |
| Name | IL-13 |
| Quantity | 10 μg |
| Source | <EM>E. coli</EM>-derived human IL-13 protein Gly21-Asn132 |
| Storage Requirements | Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution |
| Regulatory Status | RUO |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction