missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-1 alpha / IL1A / IL1F1 Protein
Recombinant Protein
Brand: enQuireBio™ QP5298-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000566.3 | |
IL-1 alpha / IL1A / IL1F1 Protein | |
Research Use Only | |
IL1A | |
Recombinant Protein | |
E. coli | |
SAPFSFLSNV KYNFMRIIKY EFILNDALNQ SIIRANDQYL TAAALHNLDE AVKFDMGAYK SSKDDAKITV ILRISKTQLY VTAQDEDQPV LLKEMPEIPK TITGSETNLL FFWETHGTKN YFTSVAHPNL FIATKQDYWV CLAGGPPSIT DFQILENQA | |
Greater than 95% as determined by SDS-PAGE. |
3552 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
To pass QC, D10S cell proliferation must be significantly induced by less than 50 pg/ml or human IL-1 alpha / IL1A / IL1F1 (or activity must be at least 2.0 x 10^7 units/mg). | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |