missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGFBP6 Protein
A cDNA sequence encoding the IGFBP6 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12383-2ug
Additional Details : Weight : 0.01000kg
Specifications
3489 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSRCPGCGQ GVQAGCPGGC VEEEDGGSPA EGCAEAEGCL RREGQECGVY TPNCAPGLQC HPPKDDEAPL RALLLGRGRC LPARAPAVAE ENPKESKPQA GTARPQDVNR RDQQRNPGTS TTPSQPNSAG VQDTEMGPCR RHLDSVLQQL QTEVYRGAQT LYVPNCDHRG FYRKRQCRSS QGQRRGPCWC VDRMGKSLPG SPDGNGSSSC PTGSSG | |
Greater than 85% as determined by SDS-PAGE. |
IGFBP6 Protein | |
Research Use Only | |
IGFBP6 | |
Human | |
His | |
The IGFBP6 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |