missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGF1 Protein
A cDNA sequence encoding the IGF1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12380-2ug
Additional Details : Weight : 0.01000kg
Specifications
3479 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
IGF1 is supplied in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |
IGF1 Protein | |
Research Use Only | |
IGF1 | |
Human | |
GST |