missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFNG / Interferon Gamma Protein
Recombinant Protein
Brand: enQuireBio™ QP5378-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000610.2 | |
IFNG / Interferon Gamma Protein | |
Research Use Only | |
IFNG | |
Recombinant Protein | |
E. coli | |
MQDPYVKEAE NLKKYFNAGH SDVADNGTLF LGILKNWKEE SDRKIMQSQI VSFYFKLFKN FKDDQSIQKS VETIKEDMNV KFFNSNKKKR DDFEKLTNYS VTDLNVQRKA IHELIQVMAE LSPAAKTGKR KRSQMLFQGR RASQ | |
Greater than 95% as determined by SDS-PAGE. |
3458 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity in the Viral CPE assay using EMC virus on A549 cells assay. The bioactivity acceptance criteria is set at 1.0 x 10^7 units/mg. | |
Human | |
Untagged | |
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5 |