missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFNG / Interferon Gamma Protein
Recombinant Protein
£80.80 - £1210.00
Specifications
Accession Number | NP_000610.2 |
---|---|
Gene ID (Entrez) | 3458 |
Name | IFNG / Interferon Gamma Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15976019
|
enQuireBio™
QP5378-20UG |
20 μg |
£80.80
20µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956019
|
enQuireBio™
QP5378-100UG |
100 μg |
£191.00
100µg |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986019
|
enQuireBio™
QP5378-250UG |
250 μg |
£421.00
250µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996019
|
enQuireBio™
QP5378-500UG |
500 μg |
£715.00
500µg |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966019
|
enQuireBio™
QP5378-1MG |
1 mg |
£1210.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_000610.2 | |
IFNG / Interferon Gamma Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity in the Viral CPE assay using EMC virus on A549 cells assay. The bioactivity acceptance criteria is set at 1.0 x 10^7 units/mg. | |
Human | |
Untagged | |
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5 |
3458 | |
Research Use Only | |
IFNG | |
Recombinant Protein | |
E. coli | |
MQDPYVKEAE NLKKYFNAGH SDVADNGTLF LGILKNWKEE SDRKIMQSQI VSFYFKLFKN FKDDQSIQKS VETIKEDMNV KFFNSNKKKR DDFEKLTNYS VTDLNVQRKA IHELIQVMAE LSPAAKTGKR KRSQMLFQGR RASQ | |
Greater than 95% as determined by SDS-PAGE. |