missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IFN a 2b Protein

A cDNA sequence encoding the IFN a 2b was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10674-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954608

  • £155.00 / 50µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

IFN a 2b Protein
Research Use Only
The specific activity was determined by the dose-dependent cytotoxicity of the human TF-1 cell line (human erythroleukemic indicator cell line) and is typically 0.02-0.08ng/ml.
Human
Untagged
The IFN-a 2b was lyophilized from 1 mg/ml in 1xPBS.
50 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Human (HEK293)
MCDLPQTHSL GSRRTLMLLA QMRRISLFSC LKDRHDFGFP QEEFGNQFQK AETIPVLHEM IQQIFNLFST KDSSAAWDET LLDKFYTELY QQLNDLEACV IQGVGVTETP LMKEDSILAV RKYFQRITLY LKEKKYSPCA WEVVRAEIMR SFSLSTNLQE SLRSKE
Greater than 95% as obsereved by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers