missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ICAM2 Protein
A cDNA sequence encoding the ICAM2 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | ICAM2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15950179
|
enQuireBio™
QP12363-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930179
|
enQuireBio™
QP12363-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940179
|
enQuireBio™
QP12363-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
ICAM2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ICAM2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSSDEKVFE VHVRPKKLAV EPKGSLEVNC STTCNQPEVG GLETSLDKIL LDEQAQWKHY LVSNISHDTV LQCHFTCSGK QESMNSNVSV YQPPRQVILT LQPTLVAVGK SFTIECRVPT VEPLDSLTLF LFRGNETLHY ETFGKAAPAP QEATATFNST ADREDGHRNF SCLAVLDLMS RGGNIFHKHS APKMLEIYEP VSDSQ | |
Greater than 90.0% as determined by SDS-PAGE. |