missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HTATIP2 Protein

A cDNA sequence encoding the HTATIP2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12350-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15940139

  • £2975.00 / 1mg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HTATIP2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAETEALSKL REDFRMQNKS VFILGASGET GRVLLKEILE QGLFSKVTLI GRRKLTFDEE AYKNVNQEVV DFEKLDDYAS AFQGHDVGFC CLGTTRGKAG AEGFVRVDRD YVLKSAELAK AGGCKHFNLL SSKGADKSSN FLYLQVKGEV EAKVEELKFD RYSVFRPGVL LCDRQESRPG EWLVRKFFGS LPDSWARGHS VPVVTVVRAM LNNVVRPRDK QMELLENKAI HDLGKAHGSL KP
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HTATIP2 protein solution (1 mg/ml) containing 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers