missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSPB9 Protein
A cDNA sequence encoding the HSPB9 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12336-1mg
Additional Details : Weight : 0.01000kg
Specifications
HSPB9 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MQRVGNTFSN ESRVASRCPS VGLAERNRVA TMPVRLLRDS PAAQEDNDHA RDGFQMKLDA HGFAPEELVV QVDGQWLMVT GQQQLDVRDP ERVSYRMSQK VHRKMLPSNL SPTAMTCCLT PSGQLWVRGQ CVALALPEAQ TGPSPRLGSL GSKASNLTR | |
Greater than 85% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The HSPB9 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 50% glycerol, 0.2M NaCl and 2mM DTT. |