missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HSPB6 Protein

A cDNA sequence encoding the HSPB6 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12333-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15970089

  • £98.39 / 2µg
Estimated Shipment: 13-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HSPB6 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HSPB6 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 30% glycerol, 1mM DTT, 1mM EDTA and 0.1mM PMSF.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMEIPVP VQPSWLRRAS APLPGLSAPG RLFDQRFGEG LLEAELAALC PTTLAPYYLR APSVALPVAQ VPTDPGHFSV LLDVKHFSPE EIAVKVVGEH VEVHARHEER PDEHGFVARE FHRRYRLPPG VDPAAVTSAL SPEGVLSIQA APASAQAPPP AAAK
Videos
SDS
Documents
Certificates
Special Offers

Special Offers