missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HSP27 Protein

A cDNA sequence encoding the HSP27 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12327-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15970069

  • £151.37 / 50µg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HSP27 Protein
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The HSP27 protein solution contains 20mM Hepes pH 7.5, 1mM DTT and 100mM KCl.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MTERRVPFSL LRGPSWDPFR DWYPHSRLFD QAFGLPRLPE EWSQWLGGSS WPGYVRPLPP AAIESPAVAA PAYSRALSRQ LSSGVSEIRH TADRWRVSLD VNHFAPDELT VKTKDGVVEI TGKHEERQDE HGYISRCFTR KYTLPPGVDP TQVSSSLSPE GTLTVEAPMP KLATQSNEIT IPVTFESRAQ LGGPEAAKSD ETAAK
Videos
SDS
Documents
Certificates
Special Offers

Special Offers