missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSF2BP Protein
A cDNA sequence encoding the HSF2BP was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | HSF2BP Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15950059
|
enQuireBio™
QP12323-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940059
|
enQuireBio™
QP12323-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930059
|
enQuireBio™
QP12323-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HSF2BP Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HSF2BP protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 30% glycerol and 2mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGEAGAA EEACRHMGTK EEFVKVRKKD LERLTTEVMQ IRDFLPRILN GEVLESFQKL KIVEKNLERK EQELEQLKMD CEHFKARLET VQADNIREKK EKLALRQQLN EAKQQLLQQA EYCTEMGAAA CTLLWGVSSS EEVVKAILGG DKALKFFSIT GQTMESFVKS LDGDVQELDS DESQFVFALA GIVTNVAAIA CGREFLVNSS RVLLDTILQL LGDLKPGQCT KLKVLMLMSL YNVSINLKGL KYISESPGFI PLLWWLLSDP DAEVCLHVLR LVQSVVLEPE VFSKSASEFR SSLPLQRILA MSKSRNPRLQ TAAQELLEDL RTLEHNV | |
Greater than 85.0% as determined by SDS-PAGE. |