missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HPCAL1 Protein

A cDNA sequence encoding the HPCAL1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12298-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15949978

  • £99.15 / 2µg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HPCAL1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGKQNSKLRP EVLQDLRENT EFTDHELQEW YKGFLKDCPT GHLTVDEFKK IYANFFPYGD ASKFAEHVFR TFDTNGDGTI DFREFIIALS VTSRGKLEQK LKWAFSMYDL DGNGYISRSE MLEIVQAIYK MVSSVMKMPE DESTPEKRTD KIFRQMDTNN DGKLSLEEFI RGAKSDPSIV RLLQCDPSSA SQF
Greater than 95.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The HPCAL1 protein solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT 0.2M NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers