missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HNMT Protein

A cDNA sequence encoding the HNMT was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12289-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959948

  • £3895.50 / 1mg
Estimated Shipment: 03-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HNMT Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The HNMT solution contains 20mM Tris-HCl pH 8 and 10% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMASS MRSLFSDHGK YVESFRRFLN HSTEHQCMQE FMDKKLPGII GRIGDTKSEI KILSIGGGAG EIDLQILSKV QAQYPGVCIN NEVVEPSAEQ IAKYKELVAK TSNLENVKFA WHKETSSEYQ SRMLEKKELQ KWDFIHMIQM LYYVKDIPAT LKFFHSLLGT NAKMLIIVVS GSSGWDKLWK KYGSRFPQDD LCQYITSDDL TQMLDNLGLK YECYDLLSTM DISDCFIDGD ENGDLLWDFL TETCNFNATA PPDLRAELGK DLQEPEFSAK KEGKVLFNNT LSFIVIEA
Videos
SDS
Documents
Certificates
Special Offers

Special Offers