missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HMGN1 Protein
A cDNA sequence encoding the HMGN1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | HMGN1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959938
|
enQuireBio™
QP12285-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939938
|
enQuireBio™
QP12285-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949938
|
enQuireBio™
QP12285-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HMGN1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HMGN1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl pH 8.0, 1mM DTT, 0.1M NaCl, 0.1mM PMSF and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MPKRKVSSAE GAAKEEPKRR SARLSAKPPA KVEAKPKKAA AKDKSSDKKV QTKGKRGAKG KQAEVANQET KEDLPAENGE TKTEESPASD EAGEKEAKSD LEHHHHHH | |
Greater than 95% as determined by SDS-PAGE. |