missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HMGN1 Protein

A cDNA sequence encoding the HMGN1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12285-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15939938

  • £155.00 / 10µg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HMGN1 Protein
Research Use Only
Recombinant Protein
E. coli
MPKRKVSSAE GAAKEEPKRR SARLSAKPPA KVEAKPKKAA AKDKSSDKKV QTKGKRGAKG KQAEVANQET KEDLPAENGE TKTEESPASD EAGEKEAKSD LEHHHHHH
Greater than 95% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HMGN1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl pH 8.0, 1mM DTT, 0.1M NaCl, 0.1mM PMSF and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers