missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Histone H3.3 Protein

A DNA sequence encoding the Homo sapiens (Human) Histone H3.3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6141-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051110

  • £1735.00 / 1mg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002098.1
Histone H3.3 Protein
Research Use Only
H3F3A
Recombinant Protein
E. coli
ARTKQTARKS TGGKAPRKQL ATKAARKSAP STGGVKKPHR YRPGTVALRE IRRYQKSTEL LIRKLPFQRL VREIAQDFKT DLRFQSAAIG ALQEASEAYL VGLFEDTNLC AIHAKRVTIM PKDIQLARRI RGERA
Greater than 90% as determined by SDS-PAGE.
3020
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers