missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Histone H2B type 1-C / E / F / G / I Protein

A DNA sequence encoding the Homo sapiens (Human) Histone H2B type 1-C/E/F/G/I, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8382-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004331

  • £1735.00 / 1mg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_003509.1
Histone H2B type 1-C / E / F / G / I Protein
Research Use Only
H2BFL
Recombinant Protein
E. coli
PEPAKSAPAP KKGSKKAVTK AQKKDGKKRK RSRKESYSVY VYKVLKQVHP DTGISSKAMG IMNSFVNDIF ERIAGEASRL AHYNKRSTIT SREIQTAVRL LLPGELAKHA VSEGTKAVTK YTSS
Greater than 90% as determined by SDS-PAGE.
3017
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers