missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HEXIM1 Protein

A cDNA sequence encoding the HEXIM1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12226-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15949758

  • £5730.00 / 1mg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HEXIM1 Protein
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMAE PFLSEYQHQP QTSNCTGAAA VQEELNPERP PGAEERVPEE DSRWQSRAFP QLGGRPGPEG EGSLESQPPP LQTQACPESS CLREGEKGQN GDDSSAGGDF PPPAEVEPTP EAELLAQPCH DSEASKLGAP AAGGEEEWGQ QQRQLGKKKH RRRPSKKKRH WKPYYKLTWE EKKKFDEKQS LRASRIRAEM FAKGQPVAPY NTTQFLMDDH DQEEPDLKTG LYSKRAAAKS DDTSDDDFME EGGEEDGGSD GMGGDGSEFL QRDFSETYER YHTESLQNMS KQELIKEYLE LEKCLSRMED ENNRLRLESK RLGGDDARVR ELELELDRLR AENLQLLTEN ELHRQQERAP LSKFGD
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HEXIM1 solution (0.5 mg/ml) containing 20mM Tris-HCl pH 8.0, 0.1M NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers