missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HEXA Protein
A cDNA sequence encoding the HEXA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12224-HIS-10ug
Additional Details : Weight : 0.01000kg
Specifications
HEXA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSTLEKNVL VVSVVTPGCN QLPTLESVEN YTLTINDDQC LLLSETVWGA LRGLETFSQL VWKSAEGTFF INKTEIEDFP RFPHRGLLLD TSRHYLPLSS ILDTLDVMAY NKLNVFHWHL VDDPSFPYES FTFPELMRKG SYNPVTHIYT AQDVKEVIEY ARLRGIRVLA EFDTPGHTLS WGPGIPGLLT PCYSGSEPSG TFGPVNPSLN NTYEFMSTFF LEVSSVFPDF YLHLGGDEVD FTCWKSNPEI QDFMRKKGFG EDFKQLESFY IQTLLDIVSS YGKGYVVWQE VFDNKVKIQP DTIIQVWRED IPVNYMKELE LVTKAGFRAL LSAPWYLNRI SYGPDWKDFY VVEPLAFEGT PEQKALVIGG EACMWGEYVD NTNLVPRLWP RAGAVAERLW SNKLTSDLTF AYERLSHFRC ELLRRGVQAQ PLNVGFCEQE FEQT | |
Greater than 85.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HEXA protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |