missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HERC5 Protein

A cDNA sequence encoding the HERC5 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12217-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15949728

  • £151.37 / 20µg
Estimated Shipment: 14-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

51191
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
HERC5
Human
His
HERC5 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
HERC5 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSFDLTVRR NHLIEDVLNQ LSQFENEDLR KELWVSFSGE IGYDLGGVKK EFFYCLFAEM IQPEYGMFMY PEGASCMWFP VKPKFEKKRY FFFGVLCGLS LFNCNVANLP FPLALFKKLL DQMPSLEDLK ELSPDLGKNL QTLLDDEGDN FEEVFYIHFN VHWDRNDTNL IPNGSSITVN QTNKRDYVSK YINYIFNDSV KAVYEEFRRG FYKMCDEDII KLFHPEELKD VIVGNTDYDW KTFEKNARYE PGYNSSHPTI VMFWKAFHKL TLEEKKKFLV FLTGTDRLQM KDLNNMKITF CCPESWNERD PIRALTCFSV LFLPKYSTME TVEEALQEAI NNNRGFG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers