missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Heme-binding protein 1

A DNA sequence encoding the Homo sapiens (Human) Heme-binding protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8272-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16003691

  • £145.46 / 10µg

Estimated Shipment
05-10-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_055135.1
Heme-binding protein 1
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
23593
Greater than 90% as determined by SDS-PAGE.
Research Use Only
HEBP2
Recombinant Protein
E. coli
AEPLQPDPGA AEDAAAQAVE TPGWKAPEDA GPQPGSYEIR HYGPAKWVST SVESMDWDSA IQTGFTKLNS YIQGKNEKEM KIKMTAPVTS YVEPGSGPFS ESTITISLYI PSEQQFDPPR PLESDVFIED RAEMTVFVRS FDGFSSAQKN QEQLLTLASI LREDGKVFDE KVYYTAGYNS PVKLLNRNNE VWLIQKNEPT KENE
Videos
Documents