missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Heat shock factor-binding protein 1
A DNA sequence encoding the Homo sapiens (Human) Heat shock factor-binding protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8320-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001528.1 | |
Heat shock factor-binding protein 1 | |
Research Use Only | |
HSBP1 | |
Recombinant Protein | |
E. coli | |
MAETDPKTVQ DLTSVVQTLL QQMQDKFQTM SDQIIGRIDD MSSRIDDLEK NIADLMTQAG VEELESENKI PATQK | |
Greater than 90% as determined by SDS-PAGE. |
3281 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |