missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Heat shock factor-binding protein 1

A DNA sequence encoding the Homo sapiens (Human) Heat shock factor-binding protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8320-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004001

  • £214.03 / 50µg

Estimated Shipment
06-10-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_001528.1
Heat shock factor-binding protein 1
50 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
3281
Greater than 90% as determined by SDS-PAGE.
Research Use Only
HSBP1
Recombinant Protein
E. coli
MAETDPKTVQ DLTSVVQTLL QQMQDKFQTM SDQIIGRIDD MSSRIDDLEK NIADLMTQAG VEELESENKI PATQK
Videos
Documents