missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GZMK Protein

A cDNA sequence encoding the GZMK was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12073-HIS-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15949278

  • £151.37 / 20µg
Estimated Shipment: 13-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

3003
Greater than 95.0% as determined by analysis by SDS-PAGE.
Research Use Only
GZMK
Human
His
GZMK protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 20% glycerol and 1mM DTT.
GZMK Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
ADPIIGGKEV SPHSRPFMAS IQYGGHHVCG GVLIDPQWVL TAAHCQYRFT KGQSPTVVLG AHSLSKNEAS KQTLEIKKFI PFSRVTSDPQ SNDIMLVKLQ TAAKLNKHVK MLHIRSKTSL RSGTKCKVTG WGATDPDSLR PSDTLREVTV TVLSRKLCNS QSYYNGDPFI TKDMVCAGDA KGQKDSCKGD SGGPLICKGV FHAIVSGGHE CGVATKPGIY TLLTKKYQTW IKSNLVPPHT NHHHHHH
Videos
SDS
Documents
Certificates
Special Offers

Special Offers