missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GYPC Protein
A cDNA sequence encoding the GYPC was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12071-1mg
Additional Details : Weight : 0.01000kg
Specifications
GYPC Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPMWSTRSP NSTAWPLSLE PDPGMASAST TMHTTTIAEP DPGMSGWPDG RMETSTPTIM HHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GYPC protein solution (1 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |