missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GTP-binding nuclear protein Ran Protein
A DNA sequence encoding the Homo sapiens (Human) GTP-binding nuclear protein Ran, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6583-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001287725.1 | |
GTP-binding nuclear protein Ran Protein | |
Research Use Only | |
RAN | |
Recombinant Protein | |
E. coli | |
AAQGEPQVQF KLVLVGDGGT GKTTFVKRHL TGEFEKKYVA TLGVEVHPLV FHTNRGPIKF NVWDTAGQEK FGGLRDGYYI QAQCAIIMFD VTSRVTYKNV PNWHRDLVRV CENIPIVLCG NKVDIKDRKV KAKSIVFHRK KNLQYYDISA KSNYNFEKPF LWLARKLIGD PNLEFVAMPA LAPPEVVMDP ALAAQYEHDL EVAQTTALPD EDDDL | |
Greater than 80% as determined by SDS-PAGE. |
5901 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |