missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GTF3C6 Protein

A cDNA sequence encoding the GTF3C6 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12067-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15939258

  • £155.00 / 10µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GTF3C6 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAAAADE RSPEDGEDEE EEEQLVLVEL SGIIDSDFLS KCENKCKVLG IDTERPILQV DSCVFAGEYE DTLGTCVIFE ENVEHADTEG NNKTVLKYKC HTMKKLSMTR TLLTEKKEGE ENIGGVEWLQ IKDNDFSYRP NMICNFLHEN EDEEVVASAP DKSLELEEEE IQMNDSSNLS CEQEKPMHLE IEDSGPLIDI PSETEGSVFM ETQMLP
Greater than 85.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
GTF3C6 protein solution (0.5 mg/ml) containing PBS buffer (pH 7.4), 10% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers