missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Grancalcin Protein
A DNA sequence encoding the Homo sapiens (Human) Grancalcin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6072-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_036330.1 | |
Grancalcin Protein | |
Research Use Only | |
GCA | |
Recombinant Protein | |
E. coli | |
MAYPGYGGGF GNFSIQVPGM QMGQPVPETG PAILLDGYSG PAYSDTYSSA GDSVYTYFSA VAGQDGEVDA EELQRCLTQS GINGTYSPFS LETCRIMIAM LDRDHTGKMG FNAFKELWAA LNAWKENFMT VDQDGSGTVE HHELRQAIGL MGYRLSPQTL TTIVKRYSKN GRIFFDDYVA CCVKLRALTD FFRKRDHLQQ GSANFIYDDF LQGTMAI | |
Greater than 80% as determined by SDS-PAGE. |
25801 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |