missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GPX3 Protein
A cDNA sequence encoding the GPX3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12044-2ug
Additional Details : Weight : 0.01000kg
Specifications
GPX3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MQSRGQEKSK MDCHGGISGT IYEYGALTID GEEYIPFKQY AGKYVLFVNV ASYCGLTGQY IELNALQEEL APFGLVILGF PCNQFGKQEP GENSEILPTL KYVRPGGGFV PNFQLFEKGD VNGEKEQKFY TFLKNSCPPT SELLGTSDRL FWEPMKVHDI RWNFEKFLVG PDGIPIMRWH HRTTVSNVKM DILSYMRRQA ALGVKRK | |
Greater than 85% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GPX3 solution contains 20mM Tris-HCl buffer (pH 7.5), 40% glycerol, 0.15M NaCl and 1mM DTT. |