missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GPHB5 Protein
A cDNA sequence encoding the GPHB5 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5065.00
Specifications
Name | GPHB5 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959168
|
enQuireBio™
QP12039-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939168
|
enQuireBio™
QP12039-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949168
|
enQuireBio™
QP12039-1MG |
1 mg |
£5065.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
GPHB5 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GPHB5 filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH 4. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASASSGNL RTFVGCAVRE FTFLAKKPGC RGLRITTDAC WGRCETWEKP ILEPPYIEAH HRVCTYNETK QVTVKLPNCA PGVDPFYTYP VAIRCDCGAC STATTECETI | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |