missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GP25L2 Protein
A DNA sequence encoding the Homo sapiens (Human) GP25L2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8481-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_059980.2 | |
GP25L2 Protein | |
Research Use Only | |
GP25L2 | |
Recombinant Protein | |
E. coli | |
FHIGETEKKC FIEEIPDETM VIGNYRTQLY DKQREEYQPA TPGLGMFVEV KDPEDKVILA RQYGSEGRFT FTSHTPGEHQ ICLHSNSTKF SLFAGGMLRV HLDIQVGEHA NDYAEIAAKD KLSELQLRVR QLVEQVEQIQ KEQNYQRWRE ERFRQTSE | |
Greater than 90% as determined by SDS-PAGE. |
54732 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |