missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GOPC Protein

A cDNA sequence encoding the GOPC was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12025-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959128

  • £98.39 / 5µg
Estimated Shipment: 07-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GOPC Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The GOPC protein solution contains 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MIRKVLLLKE DHEGLGISIT GGKEHGVPIL ISEIHPGQPA DRCGGLHVGD AILAVNGVNL RDTKHKEAVT ILSQQRGEIE FEVVYVAPEV DSDDENVEYE DESGHRYRLY LDELEGGGNP GASCKDTSGE IKVLQGFNKK AVTDTHENGD LGTASETPLD DGASKLDDLH TLYHKKSY
Videos
SDS
Documents
Certificates
Special Offers

Special Offers