missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GNG13 Protein
A cDNA sequence encoding the GNG13 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12018-100ug
Additional Details : Weight : 0.01000kg
Specifications
51764 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEEWDVP QMKKEVESLK YQLAFQREMA SKTIPELLKW IEDGIPKDPF LNPDLMKNNP WVEKGKC | |
Greater than 80.0% as determined by SDS-PAGE. |
GNG13 Protein | |
Research Use Only | |
GNG13 | |
Human | |
His | |
GNG13 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 40% glycerol, 2mM DTT and 0.1mM PMSF. |