missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GNB5 Protein

A DNA sequence encoding the Homo sapiens (Human) GNB5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6103-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16050820

  • £335.00 / 100µg

Estimated Shipment: 24-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_006569.1
GNB5 Protein
100 μg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
10681
Greater than 90% as determined by SDS-PAGE.
Research Use Only
GNB5
Recombinant Protein
E. coli
MATEGLHENE TLASLKSEAE SLKGKLEEER AKLHDVELHQ VAERVEALGQ FVMKTRRTLK GHGNKVLCMD WCKDKRRIVS SSQDGKVIVW DSFTTNKEHA VTMPCTWVMA CAYAPSGCAI ACGGLDNKCS VYPLTFDKNE NMAAKKKSVA MHTNYLSACS FTNSDMQILT ASGDGTCALW DVESGQLLQS FHGHGADVLC LDLAPSETGN TFVSGGCDKK AMVWDMRSGQ CVQAFETHES DINSVRYYPS GDAFASGSDD ATCRLYDLRA DREVAIYSKE SIIFGASSVD FSLSGRLLFA GYNDYTINVW DVLKGSRVSI LFGHENRVST LRVSPDGTAF CSGSWDHTLR VWA
Videos
SDS
Documents
Certificates