missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glycogen phosphorylase, liver form Protein
A DNA sequence encoding the Homo sapiens (Human) Glycogen phosphorylase, liver form, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6561-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001157412.1 | |
Glycogen phosphorylase, liver form Protein | |
Research Use Only | |
PYGL | |
Recombinant Protein | |
E. coli | |
AKPLTDQEKR RQISIRGIVG VENVAELKKS FNRHLHFTLV KDRNVATTRD YYFALAHTVR DHLVGRWIRT QQHYYDKCPK RVYYLSLEFY MGRTLQNTMI NLGLQNACDE AIYQLGLDIE ELEEIEEDAG LGNGGLGRLA ACFLDSMATL GLAAYGYGIR YEYGIFNQKI RDGWQVEEAD DWLRYGNPWE KSRPEFMLPV HFYGKVEHTN TGTKWIDTQV VLALPYDTPV PGYMNNTVNT MRLWSARAPN DFNLRDFNVG DYIQAVLDRN LAENISRVLY PNDNFFEGKE LRLKQEYFVV AATLQDIIRR FKASKFGSTR GAGTVFDAFP DQVAIQLNDT HPALAIPELM RIFVDIEKLP WSKAWELTQK TFAYTNHTVL PEALERWPVD LVEKLLPRHL EIIYEINQKH LDRIVALFPK DVDRLRRMSL IEEEGSKRIN MAHLCIVGSH AVNGVAKIHS DIVKTKVFKD FSELEPDKFQ NKTNGITPRR WLLLCNPGLA ELIAEKIGED YVKDLSQLTK LHSFLGDDVF LRELAKVKQE NKLKFSQFLE TEYKVKINPS SMFDVQVKRI HEYKRQLLNC LHVITMYNRI KKDPKKLFVP RTVIIGGKAA PGYHMAKMII KLITSVADVV NNDPMVGSKL KVIFLENYRV SLAEKVIPAT DLSEQISTAG TEASGTGNMK FMLNGALTIG TMDGANVEMA EEAGEENLFI FGMRIDDVAA LDKKGYEAKE YYEALPELKL VIDQIDNGFF SPKQPDLFKD IINMLFYHDR FKVFADYEAY VKCQDKVSQL YMNPKAWNTM VLKNIAASGK FSSDRTIKEY AQNIWNVEPS DLKISLSNES NKVNG | |
Greater than 90% as determined by SDS-PAGE. |
5836 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |