missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Glutaredoxin / GRX1 / GLRX Protein

A DNA sequence encoding the Homo sapiens (Human) Glutaredoxin / GRX1 / GLRX, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6093-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16050760

  • £384.00 / 100µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001112362.1
Glutaredoxin / GRX1 / GLRX Protein
Research Use Only
GLRX
Recombinant Protein
E. coli
MAQEFVNCKI QPGKVVVFIK PTCPYCRRAQ EILSQLPIKQ GLLEFVDITA TNHTNEIQDY LQQLTGARTV PRVFIGKDCI GGCSDLVSLQ QSGELLTRLK QIGALQ
Greater than 90% as determined by SDS-PAGE.
2745
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers