missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GLUD1 Protein
A cDNA sequence encoding the GLUD1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Gene ID (Entrez) | 2746 |
---|---|
Name | GLUD1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | GLUD1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959038
|
enQuireBio™
QP11995-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949038
|
enQuireBio™
QP11995-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939038
|
enQuireBio™
QP11995-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
2746 | |
Research Use Only | |
GLUD1 | |
Human | |
His | |
The GLUD1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
GLUD1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSSEAVADR EDDPNFFKMV EGFFDRGASI VEDKLVEDLR TRESEEQKRN RVRGILRIIK PCNHVLSLSF PIRRDDGSWE VIEGYRAQHS QHRTPCKGGI RYSTDVSVDE VKALASLMTY KCAVVDVPFG GAKAGVKINP KNYTDNELEK ITRRFTMELA KKGFIGPGID VPAPDMSTGE REMSWIADTY ASTIGHYDIN AHACVTGKPI SQGGIHGRIS ATGRGVFHGI ENFINEASYM SILGMTPGFG DKTFVVQGFG NVGLHSMRYL HRFGAKCIAV GESDGSIWNP DGIDPKELED FKLQHGSILG FPKAKPYEGS ILEADCDILI PAASEKQLTK SNAPRVKAKI IAEGANGPTT PEADKIFLER NIMVIPDLYL NAGGVTVSYF EWLKNLNHVS YGRLTFKYER DSNYHLLMSV QESLERKFGK HGGTIPIVPT AEFQDRISGA SEKDIVHSGL AYTMERSARQ IMRTAMKYNL GLDLRTAAYV NAIEKVFKVY NEAGVTFT | |
Greater than 80.0% as determined by SDS-PAGE. |