missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GLT8D2 Protein
A DNA sequence encoding the Homo sapiens (Human) GLT8D2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6097-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001303896.1 | |
GLT8D2 Protein | |
Research Use Only | |
GLT8D2 | |
Recombinant Protein | |
E. coli | |
KKVHKGTVPK NDADDESETP EELEEEIPVV ICAAAGRMGA TMAAINSIYS NTDANILFYV VGLRNTLTRI RKWIEHSKLR EINFKIVEFN PMVLKGKIRP DSSRPELLQP LNFVRFYLPL LIHQHEKVIY LDDDVIVQGD IQELYDTTLA LGHAAAFSDD CDLPSAQDIN RLVGLQNTYM GYLDYRKKAI KDLGISPSTC SFNPGVIVAN MTEWKHQRIT KQLEKWMQKN VEENLYSSSL GGGVATSPML IVFHGKYSTI NPLWHIRHLG WNPDARYSEH FLQEAKLLHW NGRHKPWDFP SVHNDLWESW FVPDPAGIFK LNHHS | |
Greater than 90% as determined by SDS-PAGE. |
83468 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |