missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GLIPR1 / RTVP1 Protein

A DNA sequence encoding the Homo sapiens (Human) GLIPR1 / RTVP1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6089-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959889

  • £1735.00 / 1mg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_006842.2
GLIPR1 / RTVP1 Protein
Research Use Only
GLIPR1
Recombinant Protein
E. coli
ANILPDIENE DFIKDCVRIH NKFRSEVKPT ASDMLYMTWD PALAQIAKAW ASNCQFSHNT RLKPPHKLHP NFTSLGENIW TGSVPIFSVS SAITNWYDEI QDYDFKTRIC KKVCGHYTQV VWADSYKVGC AVQFCPKVSG FDALSNGAHF ICNYGPGGNY PTWPYKRGAT CSACPNNDKC LDNLCVNRQR DQVKRYYSVV YPGWPIYPRN R
Greater than 90% as determined by SDS-PAGE.
11010
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers