missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GIMAP6 Protein
A cDNA sequence encoding the GIMAP6 was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Name | GIMAP6 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15978968
|
enQuireBio™
QP11972-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968968
|
enQuireBio™
QP11972-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958968
|
enQuireBio™
QP11972-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
GIMAP6 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GIMAP6 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 1mM DTT and 30% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEEEEYE QIPQENPPEE LSQDPVLELS GGLREKEQKT PRRLRLILMG KTGSGKSATG NSILGRDVFE SKLSTRPVTK TSQRRSREWA GKELEVIDTP NILSPQVSPE VADAICQAIV LSAPGPHAVL LVTQLGRFTD EDQQVVRRLQ EVFGVGVLGH TILVFTRKED LAGGSLEDYV RETNNQALAW LDVTLARRHC GFNNRAQGEE QEAQLRELME KVEAIMWENE GDYYSNKAYQ YTQQNFRLKE LQERQVSQGQ GSEDVPGEES WLEGLSQIQK ESEEAHRCLL GKADL | |
Greater than 85% as determined by SDS-PAGE. |