missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GHRP 2 Protein
Recombinant Protein
Brand: enQuireBio™ QP11968-25mg
Additional Details : Weight : 0.01000kg
Specifications
GHRP 2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
H-D-Ala-D-2-Nal-Ala-Trp-D-Phe-Lys-NH2 | |
Greater than 99.0% as determined by analysis by RP-HPLC. |
25 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The GHRP-2 hormone was lyophilized with no additives. |