missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GFRA3 Protein

A cDNA sequence encoding the GFRA3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11959-HIS-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15968928

  • £1320.00 / 50µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GFRA3 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSDPLPTES RLMNSCLQAR RKCQADPTCS AAYHHLDSCT SSISTPLPSE EPSVPADCLE AAQQLRNSSL IGCMCHRRMK NQVACLDIYW TVHRARSLGN YELDVSPYED TVTSKPWKMN LSKLNMLKPD SDLCLKFAML CTLNDKCDRL RKAYGEACSGPHCQRHVCLR QLLTFFEKAA EPHAQGLLLC PCAPNDRGCG ERRRNTIAPN CALPPVAPNC LELRRLCFSD PLCRSRLVDF QTHCHPMDIL GTCATEQSRC LRAYLGLIGT AMTPNFVSNV NTSVALSCTC RGSGNLQEEC EMLEGFFSHN PCLTEAIAAK MRFHSQLFSQ DWPHPTFAVMAHQNEN
Greater than 85% as determined by SDS-PAGE.
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
GFRA3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) 0.4M urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers