missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDF3 Protein
A cDNA sequence encoding the GDF3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11950-20ug
Additional Details : Weight : 0.01000kg
Specifications
9573 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS AAIPVPKLSC KNLCHRHQLF INFRDLGWHK WIIAPKGFMA NYCHGECPFS LTISLNSSNY AFMQALMHAV DPEIPQAVCI PTKLSPISML YQDNNDNVIL RHYEDMVVDECGCG | |
Greater than 90.0% as determined by SDS-PAGE. |
GDF3 Protein | |
Research Use Only | |
GDF3 | |
Human | |
His | |
The protein was lyophilized from a concentrated solution (0.5 mg/ml) containing 30mM Acetate buffer pH 4. |