missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GCGR Protein
A cDNA sequence encoding the GCGR was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11939-1mg
Additional Details : Weight : 0.01000kg
Specifications
GCGR Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MQVMDFLFEK WKLYGDQCHH NLSLLPPPTE LVCNRTFDKY SCWPDTPANT TANISCPWYL PWHHKVQHRF VFKRCGPDGQ WVRGPRGQPW RDASQCQMDG EEIEVQKEVA KMYSSFQLEH HHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GCGR filtered (0.4 microm) and lyophilized from solution in acetonitrile/0,1%TFA and 1% (w/v) trehalose. |