missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GAGE12F Protein
A cDNA sequence encoding the GAGE12F was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11923-10ug
Additional Details : Weight : 0.01000kg
Specifications
100008586; 645073; 26748; 2579 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSWRGRS TYYWPRPRRY VQPPEMIGPM RPEQFSDEVE PATPEEGEPA TQRQDPAAAQ EGEDEGASAG QGPKPEAHSQ EQGHPQTGCE CEDGPDGQEM DPPNPEEVKT PEEGEKQSQC | |
Greater than 85.0% as determined by SDS-PAGE. |
GAGE12F Protein | |
Research Use Only | |
GAGE12F | |
Human | |
His | |
GAGE12F protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 10% glycerol. |