missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fumarase Protein

A cDNA sequence encoding the Fumarase was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10621-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15964438

  • £148.40 / 50µg
Estimated Shipment: 04-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

2271
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
FH
Recombinant Protein
E. coli
MASQNSFRIE YDTFGELKVP NDKYYGAQTV RSTMNFKIGG VTERMPTPVI KAFGILKRAA AEVNQDYGLD PKIANAIMKA ADEVAEGKLN DHFPLVVWQT GSGTQTNMNV NEVISNRAIE MLGGELGSKI PVHPNDHVNK SQSSNDTFPT AMHIAAAIEV HEVLLPGLQK LHDALDAKSK EFAQIIKIGR THTQDAVPLT LGQEFSGYVQ QVKYAMTRIK AAMPRIYELA AGGTAVGTGL NTRIGFAEKV AAKVAALTGL PFVTAPNKFE ALAAHDALVE LSGAMNTTAC SLMKIANDIR FLGSGPRSGL GELILPENEP GSSIMPGKVN PTQCEAMTMV AAQVMGNHVA VTVGGSNGHF ELNVFKPMMI KNVLHSARLL GDASVSFTEN CVVGIQANTE RINKLMNESL MLVTALNPHI GYDKAAKIAK TAHKNGSTLK ETAIELGYLT AEQFDEWVKP KDMLGPK
Fumarase Protein
50 μg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is > 25 unit/mg, and is defined as the amount of enzyme that cleaves 1umole of L-Malate to Fumarate per minute at pH 7.5 at 37C.
Human
Untagged
The Fumarase protein solution (1 mg/ml) contains 20mM Tris-HCl, pH 8.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers