missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fucose-1-phosphate guanylyltransferase Protein
A DNA sequence encoding the Homo sapiens (Human) Fucose-1-phosphate guanylyltransferase, was expressed in the hosts and tags indicated.
£183.90 - £1963.71
Specifications
Accession Number | NP_001186257.2 |
---|---|
Gene ID (Entrez) | 8790 |
Name | Fucose-1-phosphate guanylyltransferase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16007701
|
enQuireBio™
QP9143-YE-100UG |
100 μg |
£467.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047701
|
enQuireBio™
QP9143-YE-500UG |
500 μg |
£1265.00
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16017701
|
enQuireBio™
QP9143-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037701
|
enQuireBio™
QP9143-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057701
|
enQuireBio™
QP9143-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027701
|
enQuireBio™
QP9143-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001186257.2 | |
Fucose-1-phosphate guanylyltransferase Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
8790 | |
Research Use Only | |
FPGT | |
Recombinant Protein | |
Yeast | |
MAAARDPPEV SLREATQRKL RRFSELRGKL VARGEFWDIV AITAADEKQE LAYNQQLSEK LKRKELPLGV QYHVFVDPAG AKIGNGGSTL CALQCLEKLY GDKWNSFTIL LIHSGGYSQR LPNASALGKI FTALPLGNPI YQMLELKLAM YIDFPLNMNP GILVTCADDI ELYSIGEFEF IRFDKPGFTA LAHPSSLTIG TTHGVFVLDP FDDLKHRDLE YRSCHRFLHK PSIEKMYQFN AVCRPGNFCQ QDFAGGDIAD LKLDSDYVYT DSLFYMDHKS AKMLLAFYEK IGTLSCEIDA YGDFLQALGP GATVEYTRNT SNVIKEESEL VEMRQRIFHL LKGTSLNVVV LNNSKFYHIG TTEEYLFYFT SDNSLKSELG LQSITFSIFP DIPECSGKTS CIIQSILDSR CSVAPGSVVE YSRLGPDVSV GENCIISGSY ILTKAALPAH SFVCSLSLKM NRCLKYATMA FGVQDNLKKS VKTLSDIKLL QFFGVCFLSC LDVWNLKVTE ELFSGNKTCL SLWTARIFPV CSSLSDSVIT SLKMLNAVKN KSAFSLNSYK LLSIEEMLIY KDVEDMITYR EQIFLEISLK SSLM | |
Greater than 90% as determined by SDS-PAGE. |